Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667595

Sigma-Aldrich

N-Boc-α-(phenylsulfonyl)benzylamine

96%

Synonym(s):

(Benzenesulfonylphenylmethyl)carbamic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO4S
Molecular Weight:
347.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

169 °C (dec.)

SMILES string

CC(C)(C)OC(=O)NC(c1ccccc1)S(=O)(=O)c2ccccc2

InChI

1S/C18H21NO4S/c1-18(2,3)23-17(20)19-16(14-10-6-4-7-11-14)24(21,22)15-12-8-5-9-13-15/h4-13,16H,1-3H3,(H,19,20)

InChI key

OUHZVJQYJUWAIE-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service