Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

667579

Sigma-Aldrich

2-tert-Butyldimethylsiloxybut-3-yne

97%

Synonym(s):

tert-Butyl-dimethyl-(methyl-prop-2-ynloxy)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20OSi
CAS Number:
Molecular Weight:
184.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.423

bp

40-42 °C/9 mbar

density

0.812 g/mL at 25 °C

SMILES string

CC(O[Si](C)(C)C(C)(C)C)C#C

InChI

1S/C10H20OSi/c1-8-9(2)11-12(6,7)10(3,4)5/h1,9H,2-7H3

InChI key

DYUHDPKTGYOFOJ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service