Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

666386

Sigma-Aldrich

3,6-Dichloropyridazine-1,2-15N2, 3,6-13C2

98 atom % 15N, 99 atom % 13C, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C2H215N2Cl2
CAS Number:
Molecular Weight:
152.95
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

97% (CP)

form

solid

mp

68 °C

mass shift

M+4

SMILES string

Cl[13c]1cc[13c](Cl)[15n][15n]1

InChI

1S/C4H2Cl2N2/c5-3-1-2-4(6)8-7-3/h1-2H/i3+1,4+1,7+1,8+1

InChI key

GUSWJGOYDXFJSI-HZENGFQRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service