Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665797

Sigma-Aldrich

(S)-(−)-3,3,3-Trifluoro-1,2-epoxypropane

97%

Synonym(s):

(S)-(−)-2-(Trifluoromethyl)oxirane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$203.00
5 G
$2,030.00

$203.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$203.00
5 G
$2,030.00

About This Item

Empirical Formula (Hill Notation):
C3H3F3O
CAS Number:
Molecular Weight:
112.05
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$203.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D <1.300

bp

25-32 °C

density

1.294 g/mL at 25 °C

functional group

ether
fluoro

storage temp.

−20°C

SMILES string

FC(F)(F)[C@@H]1CO1

InChI

1S/C3H3F3O/c4-3(5,6)2-1-7-2/h2H,1H2/t2-/m0/s1

InChI key

AQZRARFZZMGLHL-REOHCLBHSA-N

Application

Precursor to a trifluoromethyl chiral auxiliary which promotes a highly diastereoselective Simmons-Smith cyclopropanation.[1]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F

Flash Point(C)

-26 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 17, 1157-1157 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service