Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665762

Sigma-Aldrich

4-Pentylphenyl 4-methoxybenzoate

97%

Synonym(s):

4-n-Pentylphenyl p-anisate, 4-Amylphenyl 4′-methoyxbenzoate, Nematal 105

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O3
CAS Number:
Molecular Weight:
298.38
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid crystal

mp

28-42 °C

storage temp.

2-8°C

SMILES string

CCCCCc1ccc(OC(=O)c2ccc(OC)cc2)cc1

InChI

1S/C19H22O3/c1-3-4-5-6-15-7-11-18(12-8-15)22-19(20)16-9-13-17(21-2)14-10-16/h7-14H,3-6H2,1-2H3

InChI key

UISXVYOLBGBYCV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service