Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

665649

Sigma-Aldrich

2,6-Bis(trifluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3F6N
CAS Number:
Molecular Weight:
215.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

bp

82 °C/18 mmHg

mp

55-59 °C

functional group

fluoro

SMILES string

FC(F)(F)c1cccc(n1)C(F)(F)F

InChI

1S/C7H3F6N/c8-6(9,10)4-2-1-3-5(14-4)7(11,12)13/h1-3H

InChI key

YPDVFTXBESQIPJ-UHFFFAOYSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

153.0 °F

Flash Point(C)

67.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service