Skip to Content
MilliporeSigma
All Photos(1)

Documents

665479

Sigma-Aldrich

(S)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)piperidine

97%

Synonym(s):

(S)-(+)-4-Dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-yl-piperidine, (S)-PipPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22NO2P
Molecular Weight:
399.42
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

mp

206-213 °C

SMILES string

C1CCN(CC1)P2Oc3ccc4ccccc4c3-c5c(O2)ccc6ccccc56

InChI

1S/C25H22NO2P/c1-6-16-26(17-7-1)29-27-22-14-12-18-8-2-4-10-20(18)24(22)25-21-11-5-3-9-19(21)13-15-23(25)28-29/h2-5,8-15H,1,6-7,16-17H2

InChI key

ZYDGLCZCEANEHK-UHFFFAOYSA-N

Application

Asymmetric hydrogenation of N-aryl imines
Phosphoramidite ligand used in a rhodium-catalyzed enantioselective 1,4-addition of arylboronic acids to α,β-unsaturated carbonyl compounds.

Legal Information

Sold under license from DSM for research purposes only.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasa Mrsić et al.
Journal of the American Chemical Society, 131(24), 8358-8359 (2009-06-18)
Asymmetric hydrogenation of N-aryl acetophenone imines using iridium/PipPhos leads to very high enantioselectivities up to >99% depending on the presence of electron-donating substituents in the 2-, 3-, and 5-position of the aryl ring. If the substituent is 2-methoxy, the resultant
Journal of Organometallic Chemistry, 692, 428-428 (2007)

Articles

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service