Skip to Content
MilliporeSigma
All Photos(1)

Documents

665223

Sigma-Aldrich

Acetylene dicarboxylic acid-1-13C disodium salt

99 atom % 13C, 98% (CP)

Synonym(s):

2-Butynedioic acid-2-13C, Acetylenedicarboxylic acid-1-13C disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO2CC≡C13CO2Na
Molecular Weight:
159.01
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

>250 °C

mass shift

M+1

SMILES string

O=C(O[Na])C#C[13C](O[Na])=O

InChI

1S/C4H2O4.2Na/c5-3(6)1-2-4(7)8;;/h(H,5,6)(H,7,8);;/q;2*+1/p-2/i1+1;;

InChI key

DJVYWPRZTVTSGO-GOCMCNPZSA-L

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service