Skip to Content
MilliporeSigma
All Photos(3)

Documents

665126

Sigma-Aldrich

4-Bromo-2-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
215.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

180-184 °C

SMILES string

Cc1cc(Br)ccc1C(O)=O

InChI

1S/C8H7BrO2/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4H,1H3,(H,10,11)

InChI key

RVCJOGNLYVNRDN-UHFFFAOYSA-N

Application

Building block used in a preparation of anthranilic acids possessing antibacterial activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atli Thorarensen et al.
Bioorganic & medicinal chemistry letters, 17(10), 2823-2827 (2007-03-21)
In the past few years, a significant effort has been devoted by Pharmacia toward the discovery of novel antibiotics. We have recently described the identification of an anthranilic acid lead 1 and the optimization resulting in the advanced lead 2.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service