Skip to Content
MilliporeSigma
All Photos(1)

Documents

665029

Sigma-Aldrich

1-(1-Adamantyl)-3-(2,4,6-trimethylphenyl)imidazolinium chloride

Synonym(s):

1-(1-Adamantyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31ClN2
CAS Number:
Molecular Weight:
358.95
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reagent type: catalyst
reaction type: Ring-Opening Polymerization

mp

263-280 °C

SMILES string

[Cl-].Cc1cc(C)c(N2CC[N+](=C2)C34CC5CC(CC(C5)C3)C4)c(C)c1

InChI

1S/C22H31N2.ClH/c1-15-6-16(2)21(17(3)7-15)23-4-5-24(14-23)22-11-18-8-19(12-22)10-20(9-18)13-22;/h6-7,14,18-20H,4-5,8-13H2,1-3H3;1H/q+1;/p-1/t18-,19+,20-,22-;

InChI key

MJOMRTQWLPXDJQ-ZFCGJYDVSA-M

Application

Reactant for:
  • Preparation of chelated ruthenium alkoxybenzylidene imidazolidene carbene pivalate catalysts for Z-selective olefin metathesis
  • Use in ring-opening metathesis polymerization and ring-closing metathesis reactions via Grubbs catalyst mediated reactions
  • Preparation of adamantyl-substituted N-heterocyclic carbene ligands in second-generation Grubbs-type iridium metathesis catalysis

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service