Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

663492

Sigma-Aldrich

Nafion perfluorinated resin solution

greener alternative

20 wt. % in lower aliphatic alcohols and water, contains 34% water

Synonym(s):

D-2020

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203
NACRES:
NA.23

contains

34% water

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

20 wt. % in lower aliphatic alcohols and water

density

1.02 g/mL at 25 °C

greener alternative category

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Nafion perfluorinated resin solution is widely used as an electrolytic material. It can be used in the formation of a composite membrane with metal oxides. The membrane further facilitates the formation of chemically stable proton exchange membrane (PEM) fuel cells. It may be coated on mesoporous metallic films to improve the detection and selectivity of biomarkers. It may also form a composite electrode material with graphene for glucose biosensors.[1][2][3][4][5]
Nafion resin was used to produce nanometer thick Nafion layers on silane treated slides by the solution casting method.[6]
Support material for an integrated system which used solar energy to drive chemical reactions. Acts as a superacid catalyst for organic synthesis. When formed as metal composites, can be used as artificial muscle actuators, solid electrolytes for the analysis of electroactive species in organic media, or as electrodes in organic electrosynthesis.

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucose biosensor of ruthenium-doped TiO2 sensing electrode by co-sputtering system
Chou JC, et al.
Microelectronics reliability (2010)
Electrokinetic Measurements of Thin Nafion Films.
Barbati AC and Kirby BJ
Langmuir, 30(8), 1985-1993 (2014)
An enzymatic glucose biosensor based on a glassy carbon electrode modified with manganese dioxide nanowires
Zhang L, et al.
Microchimica Acta, 54-56 (2013)
Organically functionalized titanium oxide/Nafion composite proton exchange membranes for fuel cells applications
Cozzi D, et al.
Journal of Power Sources, 248, 1127-1132 (2014)
Preparation and evaluation of Nafion/SnO 2 nanocomposite for improving the chemical durability of proton exchange membranes in fuel cells
Taghizadeh MT and Vatanparast M
Royal Society of Chemistry Advances, 6(62), 56819-56826 (2016)

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service