Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662941

Sigma-Aldrich

2,5-Thiophenedicarbonyl dichloride

97%

Synonym(s):

2,5-Bis(chlorocarbonyl)thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl2O2S
CAS Number:
Molecular Weight:
209.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

150-152 °C/11 mmHg

mp

43-47 °C

storage temp.

2-8°C

SMILES string

ClC(=O)c1ccc(s1)C(Cl)=O

InChI

1S/C6H2Cl2O2S/c7-5(9)3-1-2-4(11-3)6(8)10/h1-2H

InChI key

UCRQTPGCFCBLOX-UHFFFAOYSA-N

Application

Building block for the synthesis of chiral bis-oxazoline ligands used in copper- and ruthenium-catalyzed asymmetric cyclopropanations.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 45, 5649-5649 (2004)
Synthetic Communications, 35, 2665-2665 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service