Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662836

Sigma-Aldrich

(N-Boc-β-amino)-Ala-OH

Synonym(s):

Boc-Dap-OH, (S)-2-[(tert-Butoxycarbonyl)amino]-3-aminopropionic acid, (S)-3-Amino-2-(tert-butoxycarbonyl)aminopropionic acid, (S)-3-Amino-2-(tert-butoxycarbonylamino)propanoic acid, 3-Amino-(tert-butoxycarbonyl)-L-alanine, Nα-BOC-(S)-β-aminoalanine, Nα-Boc-L-β-aminoalanine, N2-(tert-Butoxycarbonyl)-(S)-2,3-diaminopropionic acid, N2-tert-Butoxycarbonyl-L-2,3-diaminopropionic acid, Nα-Boc-L-2,3-diaminopropionic acid, Boc-Dpr-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4
CAS Number:
Molecular Weight:
204.22
Beilstein:
4182136
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

210 °C (dec.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CN)C(O)=O

InChI

1S/C8H16N2O4/c1-8(2,3)14-7(13)10-5(4-9)6(11)12/h5H,4,9H2,1-3H3,(H,10,13)(H,11,12)/t5-/m0/s1

InChI key

KRJLRVZLNABMAT-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Protein assembly directed by synthetic molecular recognition motifs
  • Solid phase synthesis of gramicidin S cyclic analogs with antibiotic and hemolytic activities
  • Synthesis of HCV protease inhibitor modified analogs
  • Solid phase synthesis of peptidic V1a receptor agonists
  • Directed peptide assembly at lipid-water interface

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service