Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

661759

Sigma-Aldrich

2-Pyridinesulfonic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO3S
CAS Number:
Molecular Weight:
159.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

244-249 °C

functional group

sulfonic acid

SMILES string

OS(=O)(=O)c1ccccn1

InChI

1S/C5H5NO3S/c7-10(8,9)5-3-1-2-4-6-5/h1-4H,(H,7,8,9)

InChI key

KZVLNAGYSAKYMG-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela Rendošová et al.
Journal of inorganic biochemistry, 186, 206-216 (2018-07-02)
In the current study the ability of silver pyridine-2-sulfonate complex to exert multiple biological activities is compared with the pharmacological action of silver sulfadiazine (AgSD). Polymeric form of {[Ag(py-2-SO3)]}n (AgPS) was synthesized and characterized by analytical techniques (IR, CHN, TG/DTA
Sebastian Faßbender et al.
Analytical and bioanalytical chemistry, 412(23), 5637-5646 (2020-07-03)
In this work, a method for species-specific isotopic analysis of sulfur via capillary electrophoresis hyphenated on-line with multicollector ICP-MS (CE/MC-ICP-MS) was developed. Correction for the mass bias caused by instrumental mass discrimination was realized via external correction with multiple-injection sample-standard

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service