Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

661724

Sigma-Aldrich

Vinyltrimethylsilane

≥99.5%

Synonym(s):

Trimethyl(vinyl)silane, (Trimethylsilyl)ethylene, Ethenyltrimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCHCH2
CAS Number:
Molecular Weight:
100.23
Beilstein:
956572
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

4.44 psi ( 20 °C)

Assay

≥99.5%

refractive index

n20/D 1.391 (lit.)

bp

55 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C=C

InChI

1S/C5H12Si/c1-5-6(2,3)4/h5H,1H2,2-4H3

InChI key

GCSJLQSCSDMKTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Preparation of silyl-ethers by Rh(I) catalysis.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-11.2 °F

Flash Point(C)

-24 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jung-Woo Park et al.
Organic letters, 9(20), 4073-4076 (2007-09-11)
Highly efficient O-silylation of alcohol with vinylsilane was developed using a catalyst system consisting of [(COE)(2)RhCl](2) and HCl. In this reaction, a key intermediate is chlorosilane, generated from vinylsilane and HCl, which can be regenerated in the catalytic cycle. Various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service