Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

661260

Sigma-Aldrich

4-(4-Methoxyphenoxy)benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3COC6H4OC6H4CHO
CAS Number:
Molecular Weight:
228.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

58-62 °C

SMILES string

[H]C(=O)c1ccc(Oc2ccc(OC)cc2)cc1

InChI

1S/C14H12O3/c1-16-12-6-8-14(9-7-12)17-13-4-2-11(10-15)3-5-13/h2-10H,1H3

InChI key

XXZKVKMXUPYUTR-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service