Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

660396

Sigma-Aldrich

4-Chloropyridine-3-carboxylic acid

96%

Synonym(s):

4-Chloronicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO2
CAS Number:
Molecular Weight:
157.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

139-143 °C

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1cnccc1Cl

InChI

1S/C6H4ClNO2/c7-5-1-2-8-3-4(5)6(9)10/h1-3H,(H,9,10)

InChI key

IMRGVWZLCZERSQ-UHFFFAOYSA-N

Application

Building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. Also used in a synthesis of heterocyclic analogues of xanthone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 4219-4219 (2006)
Tetrahedron, 61, 9102-9102 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service