Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

659711

Sigma-Aldrich

(R)-C3-TunePhos-ruthenium complex

Synonym(s):

[Chloro(R)-C3-TunePhos)(p-cymene)ruthenium(II)] chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H46Cl2O2P2Ru
CAS Number:
Molecular Weight:
900.81
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

mp

>300 °C

SMILES string

[Cl-].Cl[Ru+].C[C]1[CH][CH][C]([CH][CH]1)C(C)C.C2COc3cccc(P(c4ccccc4)c5ccccc5)c3-c6c(OC2)cccc6P(c7ccccc7)c8ccccc8

InChI

1S/C39H32O2P2.C10H14.2ClH.Ru/c1-5-16-30(17-6-1)42(31-18-7-2-8-19-31)36-26-13-24-34-38(36)39-35(41-29-15-28-40-34)25-14-27-37(39)43(32-20-9-3-10-21-32)33-22-11-4-12-23-33;1-8(2)10-6-4-9(3)5-7-10;;;/h1-14,16-27H,15,28-29H2;4-8H,1-3H3;2*1H;/q;;;;+2/p-2

InChI key

BOERIKXMDOKSHH-UHFFFAOYSA-L

Application

Provides comparable or superior enantioselectivities and catalytic abilities to BINAP in Ru-catalyzed asymmetric hydrogenations of β-keto esters, cyclic β-(acylamino)acrylates and α-phthalimide ketones.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aiwen Lei et al.
Journal of the American Chemical Society, 126(6), 1626-1627 (2004-02-12)
A new type of alpha-phthalimide ketones was hydrogenated in excellent enantioselectivity by using a Ru-(C3-TunePhos) complex as the catalyst. Up to 10 000 turnovers have been achieved in more than 99% ee in the hydrogenation reaction. A dynamic kinetic resolution
Wenjun Tang et al.
Journal of the American Chemical Society, 125(32), 9570-9571 (2003-08-09)
Hydrogenation of a series of cyclic beta-(acylamino)acrylates with tetrasubstituted olefins has been accomplished successfully with the use of Ru catalysts with chiral biaryl ligands such as C3-TunaPhos, and up to over 99% ee's have been achieved. This methodology provides an
Synthesis of chiral bisphosphines with tunable bite angles and their applications in asymmetric hydrogenation of beta-ketoesters.
Z Zhang et al.
The Journal of organic chemistry, 65(19), 6223-6226 (2000-09-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service