Skip to Content
MilliporeSigma
All Photos(1)

Documents

659665

Sigma-Aldrich

Poly(ethylene glycol)-block-polylactide methyl ether

PEG average Mn 350, PLA average Mn 1,000

Synonym(s):

PEG-b-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)x(COCHCH3O)yH
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

PEG average Mn 350
PLA average Mn 1,000

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

Biodegradable, water-soluble polymer. Applications include drug encapsulation and drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service