Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

658812

Sigma-Aldrich

Tetra-N-phenylbenzidine

97%

Synonym(s):

N,N,N′,N′-Tetraphenylbenzidine, TPB, Tetraphenylbenzidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H28N2
CAS Number:
Molecular Weight:
488.62
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

230-234 °C

SMILES string

c1ccc(cc1)N(c2ccccc2)c3ccc(cc3)-c4ccc(cc4)N(c5ccccc5)c6ccccc6

InChI

1S/C36H28N2/c1-5-13-31(14-6-1)37(32-15-7-2-8-16-32)35-25-21-29(22-26-35)30-23-27-36(28-24-30)38(33-17-9-3-10-18-33)34-19-11-4-12-20-34/h1-28H

InChI key

DCZNSJVFOQPSRV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Conjugated-polymer-functionalized graphene oxide: synthesis and nonvolatile rewritable memory effect.
Xiao-Dong Zhuang et al.
Advanced materials (Deerfield Beach, Fla.), 22(15), 1731-1735 (2010-05-25)
P Rapta et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(2), 357-362 (2000-03-22)
A newly developed in situ electron paramagnetic resonance (EPR)/ultraviolet-visible (UV-vis) spectroelectrochemical cell equipped with a laminated indium-tin oxide (ITO) working electrode was used in the investigation of various organic substrates which are potential hole-transporting materials. The experiment demonstrated the possibility
T Körzdörfer et al.
The Journal of chemical physics, 131(3), 034310-034310 (2009-07-25)
Perylene bisimide and triphenyl diamine are prototypical organic dyes frequently used in organic solar cells and light emitting devices. Recent Forster-resonant-energy-transfer experiments on a bridged organic dyad consisting of triphenyl diamine as an energy-donor and perylene bisimide as an energy-acceptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service