Skip to Content
MilliporeSigma
All Photos(1)

Documents

658588

Sigma-Aldrich

4,6-Dimethoxy-3-methylindole

97%

Synonym(s):

4,6-Dimethoxy-3-methyl-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO2
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

83-86 °C

SMILES string

COc1cc(OC)c2c(C)c[nH]c2c1

InChI

1S/C11H13NO2/c1-7-6-12-9-4-8(13-2)5-10(14-3)11(7)9/h4-6,12H,1-3H3

InChI key

JWWAWAORLJVTLE-UHFFFAOYSA-N

Application

Reactant for:
  • Acid-catalyzed reactions with ketones
  • Synthesis of pyrrolo[1,2-a]indoles
  • Synthesis of indolo[2,3-c]quinolines
  • Nitration and oxidative dimerization reactions with nitric acid

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service