Skip to Content
MilliporeSigma
All Photos(1)

Documents

657840

Sigma-Aldrich

1-(2,4-Dichlorobenzyl)piperazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14Cl2N2
CAS Number:
Molecular Weight:
245.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.571

density

1.255 g/mL at 25 °C

SMILES string

Clc1ccc(CN2CCNCC2)c(Cl)c1

InChI

1S/C11H14Cl2N2/c12-10-2-1-9(11(13)7-10)8-15-5-3-14-4-6-15/h1-2,7,14H,3-6,8H2

InChI key

VYKXBWXIOSLDQR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service