Skip to Content
MilliporeSigma
All Photos(1)

Documents

657530

Sigma-Aldrich

3-((2′-Chloro-5′-(trifluoromethyl)phenoxy)methyl)phenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11BClF3O3
CAS Number:
Molecular Weight:
330.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

form

solid

mp

212-216 °C

SMILES string

OB(O)c1cccc(COc2cc(ccc2Cl)C(F)(F)F)c1

InChI

1S/C14H11BClF3O3/c16-12-5-4-10(14(17,18)19)7-13(12)22-8-9-2-1-3-11(6-9)15(20)21/h1-7,20-21H,8H2

InChI key

IRBNKOHBJCGIFU-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service