656704
(S)-(−)-3-(Dimethylamino)pyrrolidine
97%
Synonym(s):
(3S)-N,N-Dimethyl-3-pyrrolidinamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
optical activity
[α]/D −14±2°, c = 1 in ethanol
refractive index
n20/D 1.4650 (lit.)
bp
160 °C/760 mmHg (lit.)
density
0.899 g/mL at 25 °C (lit.)
functional group
amine
storage temp.
2-8°C
SMILES string
CN(C)[C@H]1CCNC1
InChI
1S/C6H14N2/c1-8(2)6-3-4-7-5-6/h6-7H,3-5H2,1-2H3/t6-/m0/s1
InChI key
AVAWMINJNRAQFS-LURJTMIESA-N
Application
(S)-(−)-3-(Dimethylamino)pyrrolidine can be used as a reactant in the syntheses of oxadiazolylthiazole and xanthone derivatives of biological importance.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
124.1 °F
Flash Point(C)
51.15 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nonpeptidic amphiphilic xanthone derivatives: structure--activity relationship and membrane-targeting properties
Koh J-J, et al.
Journal of Medicinal Chemistry, 59(1), 171-193 (2016)
Oxadiazolylthiazoles as novel and selective antifungal agents
Hagras M, et al.
European Journal of Medicinal Chemistry, 189, 112046-112046 (2020)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service