Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655724

Sigma-Aldrich

9H-Carbazole-2-boronic acid pinacol ester

90%

Synonym(s):

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole, 9H-Carbazol-2-ylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20BNO2
CAS Number:
Molecular Weight:
293.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

SMILES string

CC1(C)OB(OC1(C)C)c2ccc3c(c2)[nH]c4ccccc34

InChI

1S/C18H20BNO2/c1-17(2)18(3,4)22-19(21-17)12-9-10-14-13-7-5-6-8-15(13)20-16(14)11-12/h5-11,20H,1-4H3

InChI key

RLSJGSFDSSYNPL-UHFFFAOYSA-N

Other Notes

Remainder is carbazole

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service