Skip to Content
MilliporeSigma
All Photos(1)

Documents

655406

Sigma-Aldrich

2-(2-Dicyclohexylphosphinophenyl)-1,3-dioxolane

97%

Synonym(s):

Dicyclohexyl(2-(1,3-dioxolan-2-yl)phenyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31O2P
CAS Number:
Molecular Weight:
346.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

82-85 °C (lit.)

functional group

phosphine

SMILES string

C1CCC(CC1)P(C2CCCCC2)c3ccccc3C4OCCO4

InChI

1S/C21H31O2P/c1-3-9-17(10-4-1)24(18-11-5-2-6-12-18)20-14-8-7-13-19(20)21-22-15-16-23-21/h7-8,13-14,17-18,21H,1-6,9-12,15-16H2

InChI key

DIFRDTMQEINUSG-UHFFFAOYSA-N

Application

  • Ligand for preparation of palladium P,O-ligand-catalyst for Suzuki cross-coupling reactions

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service