Skip to Content
MilliporeSigma
All Photos(1)

Documents

654701

Sigma-Aldrich

Methyl 3-morpholinobenzoate

97%

Synonym(s):

1-(3-Carbomethoxyphenyl)morpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO3
Molecular Weight:
221.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

42-46 °C (lit.)

SMILES string

COC(=O)c1cccc(c1)N2CCOCC2

InChI

1S/C12H15NO3/c1-15-12(14)10-3-2-4-11(9-10)13-5-7-16-8-6-13/h2-4,9H,5-8H2,1H3

InChI key

UZHOTUXOPYLESN-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service