Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

654663

Sigma-Aldrich

(S)-(+)-1-Cbz-3-pyrrolidinol

95%

Synonym(s):

(S)-1-Benzyloxycarbonyl-3-pyrrolidinol, (S)-1-Carbobenzoxy-3-pyrrolidinol, (S)-N-Z-3-Pyrrolidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO3
CAS Number:
Molecular Weight:
221.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
95%

Assay

95%

form

solid

optical activity

[α]20/D +21°, c = 1 in chloroform

mp

71-77 °C

functional group

hydroxyl
phenyl

SMILES string

O[C@H]1CCN(C1)C(=O)OCc2ccccc2

InChI

1S/C12H15NO3/c14-11-6-7-13(8-11)12(15)16-9-10-4-2-1-3-5-10/h1-5,11,14H,6-9H2/t11-/m0/s1

InChI key

MBLJFGOKYTZKMH-NSHDSACASA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service