Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

654582

Sigma-Aldrich

N-(2-Propenyl)-1H-benzotriazole-1-carbothioamide

96%

Synonym(s):

Benzotriazole-1-carbothioic acid allylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N4S
CAS Number:
Molecular Weight:
218.28
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

56-60 °C

storage temp.

2-8°C

SMILES string

C=CCNC(=S)n1nnc2ccccc12

InChI

1S/C10H10N4S/c1-2-7-11-10(15)14-9-6-4-3-5-8(9)12-13-14/h2-6H,1,7H2,(H,11,15)

InChI key

NEHKVFMFTNUABB-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Substituted thiosemicarbazides and N-hydroxythioureas by substitution reactions with hydrazines or hydroxylamines

Used as:
  • Thioacylating agent in the preparation of thioureas

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service