Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65373

Sigma-Aldrich

4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride

≥95.0% (HPLC)

Synonym(s):

Mtr chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClO3S
CAS Number:
Molecular Weight:
248.73
Beilstein:
4867665
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Product Name

4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride, ≥95.0% (HPLC)

Assay

≥95.0% (HPLC)

composition

sulfur content, 12.2-13.5%

mp

57-61 °C

application(s)

peptide synthesis

shipped in

wet ice

storage temp.

−20°C

SMILES string

COc1cc(C)c(c(C)c1C)S(Cl)(=O)=O

InChI

1S/C10H13ClO3S/c1-6-5-9(14-4)7(2)8(3)10(6)15(11,12)13/h5H,1-4H3

InChI key

DWLYVEYCCPEHLX-UHFFFAOYSA-N

Other Notes

Protecting group reagent for amines especially ω-amine in arginine

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Wakimasu et al.
Chemical & Pharmaceutical Bulletin, 30, 2766-2766 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service