Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

653675

Sigma-Aldrich

3,4-Dimethoxyphenethyl bromide

97%

Synonym(s):

2-Methoxy-4-(2-bromoethyl)anisole, 3,4-Dimethoxy-1-(2-bromoethyl)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$81.60
5 G
$328.00

$81.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$81.60
5 G
$328.00

About This Item

Linear Formula:
H3COC6H3(OCH3)CH2CH2Br
CAS Number:
Molecular Weight:
245.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$81.60


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

bp

130-131 °C/3 mmHg (lit.)

mp

52-56 °C (lit.)

functional group

bromo

SMILES string

COc1ccc(CCBr)cc1OC

InChI

1S/C10H13BrO2/c1-12-9-4-3-8(5-6-11)7-10(9)13-2/h3-4,7H,5-6H2,1-2H3

InChI key

LTAGXVMHLFYRNK-UHFFFAOYSA-N

Related Categories

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

>229.8 °F

Flash Point(C)

> 109.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Son T Nguyen et al.
Bioorganic & medicinal chemistry, 23(9), 2024-2034 (2015-03-31)
Recently we described a novel pyranopyridine inhibitor (MBX2319) of RND-type efflux pumps of the Enterobacteriaceae. MBX2319 (3,3-dimethyl-5-cyano-8-morpholino-6-(phenethylthio)-3,4-dihydro-1H-pyrano[3,4-c]pyridine) is structurally distinct from other known Gram-negative efflux pump inhibitors (EPIs), such as 1-(1-naphthylmethyl)-piperazine (NMP), phenylalanylarginine-β-naphthylamide (PAβN), D13-9001, and the pyridopyrimidine derivatives. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service