Skip to Content
MilliporeSigma
All Photos(1)

Documents

653640

Sigma-Aldrich

Fmoc-Met-OH-13C5,15N

98 atom % 13C, 98 atom % 15N, 97% (CP)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-methionine-13C5,15N, L-Methionine-13C5,15N, N-Fmoc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3S13CH213CH213CH(15NH-Fmoc)13CO2H
CAS Number:
Molecular Weight:
377.41
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % 13C
98 atom % 15N

Assay

97% (CP)

form

solid

technique(s)

bio NMR: suitable

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+6

storage temp.

2-8°C

SMILES string

[13CH3]S[13CH2][13CH2][13C@H]([15NH]C(=O)OCC1c2ccccc2-c3ccccc13)[13C](O)=O

InChI

1S/C20H21NO4S/c1-26-11-10-18(19(22)23)21-20(24)25-12-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,17-18H,10-12H2,1H3,(H,21,24)(H,22,23)/t18-/m0/s1/i1+1,10+1,11+1,18+1,19+1,21+1

InChI key

BUBGAUHBELNDEW-MXMKRAELSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service