Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

652997

Sigma-Aldrich

2,6-Dichloro-3-(trifluoromethyl)pyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl2F3N
CAS Number:
Molecular Weight:
215.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.4848
n20/D 1.4850 (lit.)

bp

194-196 °C (lit.)
194-196 °C/760 mmHg

density

2.00 g/mL at 25 °C
2.008 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccc(Cl)nc1Cl

InChI

1S/C6H2Cl2F3N/c7-4-2-1-3(5(8)12-4)6(9,10)11/h1-2H

InChI key

UPWAAFFFSGQECJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Horikawa et al.
Chemical & pharmaceutical bulletin, 49(12), 1621-1627 (2002-01-05)
An effective and practical synthetic route to methyl 2-methoxy-6-methylaminopyridine-3-carboxylate (7), the key intermediate of 5-bromo-2-methoxy-6-methylaminopyridine-3-carboxylic acid (1), from 2,6-dichloro-3-trifluoromethylpyridine (12) was undertaken. Process improvements were highlighted by regioselectivity of 12 with a nitrogen nucleophile and conversion of the 3-trifluoromethyl group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service