Skip to Content
MilliporeSigma
All Photos(1)

Documents

652067

Sigma-Aldrich

trans-5-Chloro-1-penten-1-ylboronic acid pinacol ester

97%

Synonym(s):

(E)-5-Chloro-1-penteneboronic acid pinacol ester, E-2-(5-Chloro-pent-1-enyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-(5-Chloro-pent-1-enyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-5-Chloro-1-penteneboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20BClO2
CAS Number:
Molecular Weight:
230.54
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4630

density

1.004 g/mL at 25 °C

SMILES string

[H]\C(CCCCl)=C(\[H])B1OC(C)(C)C(C)(C)O1

InChI

1S/C11H20BClO2/c1-10(2)11(3,4)15-12(14-10)8-6-5-7-9-13/h6,8H,5,7,9H2,1-4H3/b8-6+

InChI key

FYHBHADHHWPOFL-SOFGYWHQSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service