Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

651478

Sigma-Aldrich

Gallium antimonide

(single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm

Synonym(s):

Gallium antimony, Gallium monoantimonide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaSb
CAS Number:
Molecular Weight:
191.48
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

(single crystal substrate)

resistivity

~0.1 Ω-cm

diam. × thickness

2 in. × 0.5 mm

mp

710 °C
980 °C (lit.)

density

5.619 g/mL at 25 °C
5.62 g/mL at 25 °C (lit.)

semiconductor properties

<100>

SMILES string

[Ga]#[Sb]

InChI

1S/Ga.Sb

InChI key

VTGARNNDLOTBET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

Thermal expansion: 6.1 x 10-6/oK; Thermal conductivity: 270 mW/cm.k at 300K; Mobility 600~800 cm2/V ⋅ sec
Undoped, P-type semiconductor, growth technique: LEC, carrier concentration = 1-2 × 10-17 cm-3, EPD <103 cm-2

Physical form

cubic (a = 6.095Å)

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service