Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

651168

Sigma-Aldrich

(3-Cyanophenyl)boronic acid, neopentyl glycol ester

95%

Synonym(s):

2-(3-Cyanophenyl)-5,5′-dimethyl-1,3,2-dioxaborinane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O2BN
CAS Number:
Molecular Weight:
215.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
To inquire about this product 651168, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

95%

form

solid

mp

85-87 °C

functional group

nitrile

SMILES string

CC1(C)COB(OC1)c2cccc(c2)C#N

InChI

1S/C12H14BNO2/c1-12(2)8-15-13(16-9-12)11-5-3-4-10(6-11)7-14/h3-6H,8-9H2,1-2H3

InChI key

RTYUBKQTFNAFQC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service