Skip to Content
MilliporeSigma
All Photos(1)

Documents

651044

Sigma-Aldrich

4-(4′-Chlorobenzyloxy)phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12BClO3
CAS Number:
Molecular Weight:
262.50
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

SMILES string

OB(O)c1ccc(OCc2ccc(Cl)cc2)cc1

InChI

1S/C13H12BClO3/c15-12-5-1-10(2-6-12)9-18-13-7-3-11(4-8-13)14(16)17/h1-8,16-17H,9H2

InChI key

RONZDKNBNIRWIV-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service