Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

650811

Sigma-Aldrich

1-Boc-3-piperidone

97%

Synonym(s):

tert-Butyl 3-oxopiperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

35-40 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCCC(=O)C1

InChI

1S/C10H17NO3/c1-10(2,3)14-9(13)11-6-4-5-8(12)7-11/h4-7H2,1-3H3

InChI key

RIFXIGDBUBXKEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Boc-3-piperidinone is useful in stereocontrolled synthesis of chiral compounds.

Caution

Handle under glove bag/N2.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service