Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

650242

Sigma-Aldrich

1-(2-Fluorobenzyl)piperazine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15FN2
CAS Number:
Molecular Weight:
194.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5330 (lit.)

bp

93-96 °C/0.2 mmHg (lit.)

density

1.170 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1CN2CCNCC2

InChI

1S/C11H15FN2/c12-11-4-2-1-3-10(11)9-14-7-5-13-6-8-14/h1-4,13H,5-9H2

InChI key

IGVNZJBYRPULAI-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service