Skip to Content
MilliporeSigma
All Photos(1)

Documents

650218

Sigma-Aldrich

1-(4-Chlorobenzyl)piperazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15ClN2
CAS Number:
Molecular Weight:
210.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.5580 (lit.)

bp

104-106 °C/0.1 mmHg (lit.)

density

1.140 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(CN2CCNCC2)cc1

InChI

1S/C11H15ClN2/c12-11-3-1-10(2-4-11)9-14-7-5-13-6-8-14/h1-4,13H,5-9H2

InChI key

GSJXJZOWHSTWOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Hashimoto et al.
Brain research, 590(1-2), 341-344 (1992-09-11)
The neurotoxicity of 3,4-methylenedioxymethamphetamine (MDMA) in rat brain was attenuated significantly by coadministration of several benzylpiperazines (p-nitrobenzylpiperazine, p-chlorobenzylpiperazine and 1-piperonylpiperazine), which were weak inhibitors for [3H]6-nitroquipazine binding to the 5-hydroxytryptamine (5-HT) transporter in rat brain. These results suggest that these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service