Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

649759

Sigma-Aldrich

2-Chloro-1,1,1-triethoxyethane

97%

Synonym(s):

Triethyl 2-chloroorthoacetate, 2-Chloro-1,1,1-triethoxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17ClO3
CAS Number:
Molecular Weight:
196.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.422

bp

75-80 °C/15 mmHg

density

1.031 g/mL at 20 °C (lit.)
1.024 g/mL at 25 °C

functional group

chloro
ether

SMILES string

CCOC(CCl)(OCC)OCC

InChI

1S/C8H17ClO3/c1-4-10-8(7-9,11-5-2)12-6-3/h4-7H2,1-3H3

InChI key

URFKLQSFBXBOQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service