Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648817

Sigma-Aldrich

6-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester

Synonym(s):

6-Methyl-2-pyridineboronic acid N-phenyldiethanolamine ester, 6-Methyl-2-pyridineboronic acid[2,2¢-(phenylimino)diethanol]ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19BN2O2
CAS Number:
Molecular Weight:
282.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

SMILES string

CC1=CC=CC(B(OCC2)OCCN2C3=CC=CC=C3)=N1

InChI

1S/C16H19BN2O2/c1-14-6-5-9-16(18-14)17-20-12-10-19(11-13-21-17)15-7-3-2-4-8-15/h2-9H,10-13H2,1H3

InChI key

QSRALKLINGVTIE-UHFFFAOYSA-N

Application

May contain varying amounts of Isopropanol and N-phenyldiethanolamine as stabilizers.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service