Skip to Content
MilliporeSigma
All Photos(1)

Documents

648574

Sigma-Aldrich

Sulfur-34S

99 atom % 34S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
34S
CAS Number:
Molecular Weight:
33.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

isotopic purity

99 atom % 34S

Quality Level

form

solid

mp

119 °C (lit.)

mass shift

M+2

SMILES string

[34S]

InChI

1S/S/i1+2

InChI key

NINIDFKCEFEMDL-NJFSPNSNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sulfur-34S, isotopic analogue of sulfur atom.

Application

Sulfur-34S, stable form is used to study the origin and detection of sulphur atoms in geochemistry. It acts as the reactant in the synthesis of 34S labeled gypsum (Ca34SO4 .2H2O) and used as a tracer to investigate sulfur uptake and distribution in wheat.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service