Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648507

Sigma-Aldrich

5-Chloro-8-quinolinetrifluoromethanesulfonate

97%

Synonym(s):

5-Chloro-8-quinolinetriflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5ClF3NO3S
CAS Number:
Molecular Weight:
311.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

79-83 °C (lit.)

SMILES string

FC(F)(F)S(=O)(=O)Oc1ccc(Cl)c2cccnc12

InChI

1S/C10H5ClF3NO3S/c11-7-3-4-8(9-6(7)2-1-5-15-9)18-19(16,17)10(12,13)14/h1-5H

InChI key

ICTAEAXUAWQHSL-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service