Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

647241

Sigma-Aldrich

(R)-(−)-1-Methyl-3-pyrrolidinol

97%

Synonym(s):

(3R)-1-Methylpyrrolidin-3-ol, (R)-(−)-3-Hydroxy-N-methylpyrrolidine, (R)-N-Methyl-3-pyrrolidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D -7°, c = 1% in chloroform

refractive index

n20/D 1.4640 (lit.)

bp

50-52 °C/1 mmHg (lit.)

density

0.921 g/mL at 25 °C (lit.)

SMILES string

CN1CC[C@@H](O)C1

InChI

1S/C5H11NO/c1-6-3-2-5(7)4-6/h5,7H,2-4H2,1H3/t5-/m1/s1

InChI key

FLVFPAIGVBQGET-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Asymmetric synthesis of constrained (-)-S-adenosyl-L-homocysteine (SAH) analogs as DNA methyltransferase inhibitors
  • Preparation of diaryl acylaminopyrimidines as adenosine A2A antagonists
  • Synthesis of analogs of istaroxime, a potent inhibitor of Na+,K+-ATPase
  • Synthesis of pharmacologically active stereoisomers of N-substituted soft anticholinergics

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service