Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

646571

Sigma-Aldrich

Poly(2,5-di(3,7-dimethyloctyloxy)cyanoterephthalylidene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C58H90N2O4)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

semiconductor properties

N-type (mobility~10−5 cm2/V·s)

General description

Light emitting conjugated polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Materials Design Concepts for Efficient Blue OLEDs: A Joint Theoretical and Experimental Study

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service