Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

645931

Sigma-Aldrich

4-[(4-Chloro-3-methylphenoxy)methyl]phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14BClO3
CAS Number:
Molecular Weight:
276.52
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

SMILES string

Cc1cc(OCc2ccc(cc2)B(O)O)ccc1Cl

InChI

1S/C14H14BClO3/c1-10-8-13(6-7-14(10)16)19-9-11-2-4-12(5-3-11)15(17)18/h2-8,17-18H,9H2,1H3

InChI key

OUKUADBAEFEIGP-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service