Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

644129

Sigma-Aldrich

Magnesium di-tert-butoxide

≥85%

Synonym(s):

Magnesium tert-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COMgOC(CH3)3
CAS Number:
Molecular Weight:
170.53
Beilstein:
3979348
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

≥85%

form

solid

SMILES string

CC(C)(C)O[Mg]OC(C)(C)C

InChI

1S/2C4H9O.Mg/c2*1-4(2,3)5;/h2*1-3H3;/q2*-1;+2

InChI key

RTKCPZYOLXPARI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service