Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

643823

Sigma-Aldrich

HEPES-d18

98 atom % D, 98% (CP)

Synonym(s):

4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid-d18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D18N2O4S
Molecular Weight:
256.42
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

technique(s)

NMR: suitable
bio NMR: suitable

mass shift

M+18

SMILES string

[2H]OC([2H])([2H])C([2H])([2H])N1C([2H])([2H])C([2H])([2H])N(C([2H])([2H])C1([2H])[2H])C([2H])([2H])C([2H])([2H])S(=O)(=O)O[2H]

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,11D/hD

InChI key

JKMHFZQWWAIEOD-MMODJCGESA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
696633687022655392
HEPES-d18 98 atom % D, 98% (CP)

643823

HEPES-d18

PIPES-d18 ≥98 atom % D, ≥98% (CP)

696633

PIPES-d18

MES-d13 98 atom % D, 98% (CP)

687022

MES-d13

technique(s)

NMR: suitable, bio NMR: suitable

technique(s)

bio NMR: suitable

technique(s)

NMR: suitable, bio NMR: suitable

technique(s)

NMR: suitable, bio NMR: suitable

isotopic purity

98 atom % D

isotopic purity

≥98 atom % D

isotopic purity

98 atom % D

isotopic purity

98 atom % D

assay

98% (CP)

assay

≥98% (CP)

assay

98% (CP)

assay

98% (CP)

form

solid

form

solid

form

solid

form

solid

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service