Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

642827

Sigma-Aldrich

2-Amino-5-bromobenzonitrile

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Br)CN
CAS Number:
Molecular Weight:
197.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

96-100 °C (lit.)

SMILES string

Nc1ccc(Br)cc1C#N

InChI

1S/C7H5BrN2/c8-6-1-2-7(10)5(3-6)4-9/h1-3H,10H2

InChI key

OATYCBHROMXWJO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayin Hu et al.
ChemSusChem, 11(24), 4219-4225 (2018-11-16)
The efficient transformation of CO2 into value-added chemicals with green, abundant, and cheap catalysts is an interesting and challenging topic in both green and sustainable chemistry. In this study, a series of salt-lake brines were used for the first time

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service